2-Isopropylanisole ≥98%
Supplier: Thermo Scientific
Warning
|
H53383.06
H53383.14
H53383.22
ALFAH53383.06EA
61.4
SGD
ALFAH53383.06
ALFAH53383.14
ALFAH53383.22
2-Isopropylanisole ≥98%
2-Isopropylanisole
Purity: 98%
CAS No.: 2944-47-0
Molecular Formula: C10H14O
Formula Weight: 150.22
Flash Point: 86°(187°F)
Refractive Index: 1.508
Boiling Point: 198-200°
MDL No.: MFCD00798590
Formula:
C₁₀H₁₄O MW: 150,22 g/mol Boiling Pt: 198…200 °C Density: 0,936 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00798590 CAS Number: 2944-47-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...