N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5% for applications requiring low water content, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

AS304-0100 AS304-1
BJAAAS304-0100EA 815 SGD
BJAAAS304-0100 BJAAAS304-1
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5% for applications requiring low water content, Burdick & Jackson™
N-Methyl-2-pyrrolidone (NMP)

Septum-sealed bottle


  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

Formula: C₅H₉NO
MW: 99,13 g/mol
Boiling Pt: 202 °C
Melting Pt: -24 °C
Density: 1,0259 g/cm3
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
GC Analysis 99.5 % min.
Water by Karl Fischer Titration 50 ppm max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR