2,3-Dichlorobenzenesulphonamide ≥97%
Supplier: Thermo Scientific
|
Warning
|
H32489.03
H32489.09
ALFAH32489.03EA
0
SGD
ALFAH32489.03
ALFAH32489.09
2,3-Dichlorobenzenesulphonamide ≥97%
2,3-Dichlorobenzenesulphonamide
Purity: 97%
Molecular Formula: C6H5Cl2NO2S
Formula Weight: 226.08
Melting Point: 218-221°
MDL No.: MFCD00464303
Formula:
C₆H₅Cl₂NO₂S MW: 226,08 g/mol Melting Pt: 218…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00464303 CAS Number: 82967-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...