2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98%

Supplier: Thermo Scientific
Warning

H25958.14 H25958.06 H25958.03
ALFAH25958.14EA 0 SGD
ALFAH25958.14 ALFAH25958.06 ALFAH25958.03
2,3-Difluoro-5-(trifluoromethyl)pyridine ≥98%
2,3-Difluoro-5-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 89402-42-6 Molecular Formula: C6H2F5N Formula Weight: 183.08 Flash Point: 100°(212°F) Boiling Point: 104° MDL No.: MFCD04972837

Formula: C₆H₂F₅N
MW: 183,08 g/mol
Boiling Pt: 104 °C
Density: 1,47
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD04972837
CAS Number: 89402-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR