2,4-Difluorothiobenzamide ≥97%

Supplier: Thermo Scientific
Danger

H26758.03 H26758.06
ALFAH26758.03EA 0 SGD
ALFAH26758.03 ALFAH26758.06
2,4-Difluorothiobenzamide ≥97%
2,4-Difluorothiobenzamide

Purity: 97% CAS No.: 175276-92-3 Molecular Formula: C7H5F2NS Formula Weight: 173.18 Melting Point: 128-132° UN No.: 2811 MDL No.: MFCD00085011

Formula: C₇H₅F₂NS
MW: 173,19 g/mol
Melting Pt: 128…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00085011
CAS Number: 175276-92-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR