2,5-Bis(trifluoromethyl)benzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2,5-Bis(trifluoromethyl)benzene-1-sulphonyl chloride

L19878.03 L19878.06
ALFAL19878.03EA 0 SGD
ALFAL19878.03 ALFAL19878.06
2,5-Bis(trifluoromethyl)benzenesulphonyl chloride ≥97%
2,5-Bis(trifluoromethyl)benzenesulphonyl chloride

Purity: 97% CAS No.: 351003-22-0 Molecular Formula: C8H3ClF6O2S Formula Weight: 312.61 Melting Point: 59-63° UN No.: 3261 MDL No.: MFCD03094042

Formula: C₈H₃ClF₆O₂S
MW: 312,62 g/mol
Melting Pt: 59…63 °C
Storage Temperature: Ambient
MDL Number: MFCD03094042
CAS Number: 351003-22-0
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR