2,5-Difluorothiobenzamide ≥97%

Supplier: Thermo Scientific
Danger

H32225.03 H32225.06
ALFAH32225.03EA 0 SGD
ALFAH32225.03 ALFAH32225.06
2,5-Difluorothiobenzamide ≥97%
2,5-Difluorothiobenzamide

Purity: 97% Molecular Formula: C7H5F2NS Formula Weight: 173.18 Melting Point: 109-113° UN No.: 2811 MDL No.: MFCD09757552

Formula: C₇H₅F₂NS
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD09757552
CAS Number: 1029623-72-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR