2,6-Dichloro-4-hydroxybenzaldehyde ≥98%
Supplier: Thermo Scientific
|
Warning
|
H28395.06
H28395.03
ALFAH28395.06EA
0
SGD
ALFAH28395.06
ALFAH28395.03
2,6-Dichloro-4-hydroxybenzaldehyde ≥98%
2,6-Dichloro-4-hydroxybenzaldehyde
Purity: 98%
CAS No.: 60964-09-2
Molecular Formula: C7H4Cl2O2
Formula Weight: 191.01
Melting Point: 224-229°
MDL No.: MFCD06797919
Formula:
C₇H₄Cl₂O₂ Melting Pt: 224…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797919 CAS Number: 60964-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...