2-Amino-5-nitro-3-picoline 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Amino-3-methyl-5-nitropyridine, 3-Methyl-5-nitro-2-pyridylamine
L19844.03
L19844.06
L19844.14
ALFAL19844.03EA
0
SGD
ALFAL19844.03
ALFAL19844.06
ALFAL19844.14
2-Amino-5-nitro-3-picoline 97%
2-Amino-5-nitro-3-picoline
Purity: 97%
CAS No.: 18344-51-9
Molecular Formula: C6H7N3O2
Formula Weight: 153.14
Melting Point: 258-261°
Beilstein Reference: 126947
Formula:
C₆H₇N₃O₂ MW: 153,14 g/mol Melting Pt: 258…261 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095060 CAS Number: 18344-51-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...