2-Chlorobenzimidazole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Chloro-1H-benzo[d]imidazole, 2-Chloro-1H-benzimidazole

A12193.06 A12193.14 A12193.22
ALFAA12193.06EA 197 SGD
ALFAA12193.06 ALFAA12193.14 ALFAA12193.22
2-Chlorobenzimidazole ≥97%
2-Chlorobenzimidazole

Purity: 97% CAS No.: 4857-06-1 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Melting Point: ca 180-185° Beilstein Reference: 116526

Formula: C₇H₅ClN₂
MW: 152,58 g/mol
Melting Pt: ∼ 180…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00051944
CAS Number: 4857-06-1
EINECS: 225-453-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR