2-Fluoro-6-(trifluoromethyl)pyridine ≥98%
Supplier: Thermo Scientific
|
Warning
|
H32633.03
H32633.06
ALFAH32633.03EA
0
SGD
ALFAH32633.03
ALFAH32633.06
2-Fluoro-6-(trifluoromethyl)pyridine ≥98%
2-Fluoro-6-(trifluoromethyl)pyridine
Purity: 98%
CAS No.: 94239-04-0
Molecular Formula: C6H3F4N
Formula Weight: 165.09
UN No.: 1993
MDL No.: MFCD03092901
Formula:
C₆H₃F₄N MW: 165,09 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03092901 CAS Number: 94239-04-0 EINECS: 428-100-3 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...