2-Hydroxy-5-nitro-3-picoline ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Methyl-5-nitropyridin-2-ol, 3-Methyl-5-nitropyridin-2(1H)-one, 3-Methyl-5-nitro-2-pyridone, 2-Hydroxy-3-methyl-5-nitropyridine

L19866.03 L19866.06
ALFAL19866.03EA 0 SGD
ALFAL19866.03 ALFAL19866.06
2-Hydroxy-5-nitro-3-picoline ≥97%
2-Hydroxy-5-nitro-3-picoline

Purity: 97% CAS No.: 21901-34-8 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Melting Point: 228-231° Beilstein Reference: 126949

Formula: C₆H₆N₂O₃
MW: 154,13 g/mol
Melting Pt: 228…231 °C
Storage Temperature: Ambient
MDL Number: MFCD03095073
CAS Number: 21901-34-8
EINECS: 244-647-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR