2-Iodoimidazole 96%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Iodo-1H-imidazole

H51868.MD H51868.03
ALFAH51868.MDEA 144 SGD
ALFAH51868.MD ALFAH51868.03
2-Iodoimidazole 96%
2-Iodoimidazole

Purity: 96% CAS No.: 3034-62-6 Molecular Formula: C3H3IN2 Formula Weight: 193.96 Melting Point: 190-191° MDL No.: MFCD00159701

Formula: C₃H₃IN₂
MW: 193,97 g/mol
Melting Pt: 190…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00159701
CAS Number: 3034-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR