2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Nitro-4-(trifluoromethyl)benzene-1-sulphonyl chloride
L19238.03
L19238.06
ALFAL19238.03EA
0
SGD
ALFAL19238.03
ALFAL19238.06
2-Nitro-4-(trifluoromethyl)benzenesulphonyl chloride ≥98%
2-Nitro-5-(trifluoromethyl)benzenesulphonyl chloride
Purity: 98%
CAS No.: 837-95-6
Molecular Formula: C7H3ClF3NO4S
Formula Weight: 289.62
Flash Point:
Formula:
C₇H₃ClF₃NO₄S MW: 289,62 g/mol Boiling Pt: 325 °C (1013 hPa) Melting Pt: 42…44 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013456 CAS Number: 837-95-6 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...