3,5-Dibromobenzamide ≥97%

Supplier: Thermo Scientific
Warning

B24946.03 B24946.06 B24946.14
ALFAB24946.03EA 0 SGD
ALFAB24946.03 ALFAB24946.06 ALFAB24946.14
3,5-Dibromobenzamide ≥97%
3,5-Dibromobenzamide

Purity: 97% CAS No.: 175205-85-3 Molecular Formula: C7H5Br2NO Formula Weight: 278.93 Melting Point: 180-184° MDL No.: MFCD00178771

Formula: C₇H₅Br₂NO
MW: 278,93 g/mol
Melting Pt: 180…184 °C
Storage Temperature: Ambient
MDL Number: MFCD00178771
CAS Number: 175205-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR