3,5-Difluorothiobenzamide ≥97%
Supplier: Thermo Scientific
|
Danger
|
H31719.03
H31719.06
ALFAH31719.03EA
0
SGD
ALFAH31719.03
ALFAH31719.06
3,5-Difluorothiobenzamide ≥97%
3,5-Difluorothiobenzamide
Purity: 97%
CAS No.: 874791-10-3
Molecular Formula: C7H5F2NS
Formula Weight: 173.18
Melting Point: 135-139°
UN No.: 2811
MDL No.: MFCD04036082
Formula:
C₇H₅F₂NS Melting Pt: 135…139 °C Storage Temperature: Ambient |
MDL Number:
MFCD04036082 CAS Number: 874791-10-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...