3-Bromo-5-ethoxy-4-hydroxybenzaldehyde 97

Supplier: Thermo Fisher Scientific
Warning

A19925.03 A19925.06
ALFAA19925.03EA 0 SGD
ALFAA19925.03 ALFAA19925.06
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde 97
3-Bromo-5-ethoxy-4-hydroxybenzaldehyde

Purity: 97% CAS No.: 3111-37-3 Molecular Formula: C9H9BrO3 Formula Weight: 245.08 Melting Point: 141-143° MDL No.: MFCD00173904

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Melting Pt: 141…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00173904
CAS Number: 3111-37-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR