4-(Trifluoromethyl)thiobenzamide ≥97%
Supplier: Thermo Scientific
|
Danger
|
A14600.06
A14600.03
ALFAA14600.06EA
0
SGD
ALFAA14600.06
ALFAA14600.03
4-(Trifluoromethyl)thiobenzamide ≥97%
4-(Trifluoromethyl)thiobenzamide
Purity: 97%
CAS No.: 72505-21-6
Molecular Formula: C8H6F3NS
Formula Weight: 205.20
Melting Point: 135-138°
Beilstein Reference: 4672885
UN No.: 2811
MDL No.: MFCD00051806
Formula:
C₈H₆F₃NS MW: 205,2 g/mol Boiling Pt: 254 °C (1013 hPa) Melting Pt: 135…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051806 CAS Number: 72505-21-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...