4,5-Dichloro-2-methylimidazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4,5-Dichloro-2-methyl-1H-imidazole
B20667.03
B20667.06
B20667.14
ALFAB20667.03EA
0
SGD
ALFAB20667.03
ALFAB20667.06
ALFAB20667.14
4,5-Dichloro-2-methylimidazole ≥97%
4,5-Dichloro-2-methylimidazole
Purity: 97%
CAS No.: 15965-33-0
Molecular Formula: C4H4Cl2N2
Formula Weight: 151.00
Melting Point: ca 248° dec.
Beilstein Reference: 606442
Formula:
C₄H₄Cl₂N₂ MW: 150,99 g/mol Melting Pt: ∼ 248 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051639 CAS Number: 15965-33-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...