4-Bromo-2,6-difluorobenzaldehyde 95%

Supplier: Thermo Scientific
Danger

H27737.MD H27737.03
ALFAH27737.MDEA 0 SGD
ALFAH27737.MD ALFAH27737.03
4-Bromo-2,6-difluorobenzaldehyde 95%
4-Bromo-2,6-difluorobenzaldehyde

Purity: 95% CAS No.: 537013-51-7 Molecular Formula: C7H3BrF2O Formula Weight: 221.00 Melting Point: 76-79° MDL No.: MFCD03094459

Formula: C₇H₃BrF₂O
MW: 221 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD03094459
CAS Number: 537013-51-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR