4-Bromo-2-chlorobenzenesulphonamide ≥97%

Supplier: Thermo Fisher Scientific
Danger

L19719.03 L19719.06
ALFAL19719.03EA 0 SGD
ALFAL19719.03 ALFAL19719.06
4-Bromo-2-chlorobenzenesulphonamide ≥97%
4-Bromo-2-chlorobenzenesulphonamide

Purity: 97% CAS No.: 351003-59-3 Molecular Formula: C6H5BrClNO2S Formula Weight: 270.53 Melting Point: 172-176° MDL No.: MFCD03427209

Formula: C₆H₅BrClNO₂S
MW: 270.53
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD03427209
CAS Number: 351003-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR