4-Fluoro-3-nitrobenzaldehyde ≥98%

Supplier: Thermo Scientific
Warning

H26181.03 H26181.06
ALFAH26181.03EA 0 SGD
ALFAH26181.03 ALFAH26181.06
4-Fluoro-3-nitrobenzaldehyde ≥98%
4-Fluoro-3-nitrobenzaldehyde

Purity: 98% CAS No.: 42564-51-2 Molecular Formula: C7H4FO3 Formula Weight: 169.11 Flash Point: 110°(230°F) Melting Point: 44-47° MDL No.: MFCD01861388

Formula: C₇H₄FNO₃
MW: 169,11 g/mol
Melting Pt: 44…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR