4-Iodo-3-nitrobenzoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
B25626.06
B25626.14
ALFAB25626.06EA
0
SGD
ALFAB25626.06
ALFAB25626.14
4-Iodo-3-nitrobenzoic acid ≥97%
4-Iodo-3-nitrobenzoic acid
Purity: 97%
CAS No.: 35674-27-2
Molecular Formula: C7H4INO4
Formula Weight: 293.02
Melting Point: 208-211°
MDL No.: MFCD00502399
Formula:
C₇H₄INO₄ MW: 293,02 g/mol Melting Pt: 208…211 °C Storage Temperature: Ambient |
MDL Number:
MFCD00502399 CAS Number: 35674-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...