2',6-Dihydroxyflavone ≥97%

Supplier: Thermo Fisher Scientific
Warning

H27229.03 H27229.06
ALFAH27229.03EA 0 SGD
ALFAH27229.03 ALFAH27229.06
2',6-Dihydroxyflavone ≥97%
2',6-Dihydroxyflavone

Purity: 97% CAS No.: 92439-20-8 Molecular Formula: C15H10O4 Formula Weight: 254.24 MDL No.: MFCD00017676

Formula: C₁₅H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00017676
CAS Number: 92439-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR