N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Acetic acid dimethylamide, Dimethyl Acetamide, DMAC

NP071-20 071-4
BJAANP071-20EA 4660 SGD
BJAANP071-20 BJAA071-4
N,N-Dimethylacetamide ≥99.5%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
N,N-Dimethylacetamide

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 166 °C
Melting Pt: –20 °C
Density: 0.937 g/cm³ (25 °C)
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
UN: 2810
Merck Index: 13,03254
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GC Analysis 99.5 % min.
GC Impurities Eluting Before Solvent 0.5 mg/L max.
UV Absorbance @ 275 nm 0.3 AU max.
UV Absorbance @ 300 nm 0.08 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Absorbance @ 400 nm 0.005 AU max.
UV Cutoff 268 nm max.
Water by Karl Fischer Titration 0.03 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR