Diethyl ether ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

PP108-19 PP108-204
BJAAPP108-19EA 1680 SGD
BJAAPP108-19 BJAAPP108-204
Diethyl ether ≥99.9% stabilised, Purified Plus™ ACS for organic synthesis, for preparative liquid chromatography, Burdick & Jackson™
Diethyl ether

  • Low residue
  • Purified solvent
  • ACS General Use certified

Formula: (CH₃CH₂)₂O
MW: 74,12 g/mol
Boiling Pt: 35 °C
Melting Pt: -117,4 °C
Density: 0,713 g/cm³ (20 °C)
Flash Pt: -45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=9e4ec8d25df2fecee88827e09071c0f955486274&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Alcohol (as CH3CH2OH) Passes test
Assay by GC Analysis 99.9 % min.
Carbonyl (as HCHO) 0.001 % max.
Color (APHA) 10 max.
Peroxides (as H₂O₂) at time of packaging 1 ppm max.
Refractive Index (20ºC) 1.3518 - 1.3538
Residue after Evaporation 5 mg/L max.
Titrable Acid 0.2 µeq/g max.
Water by Karl Fischer Titration 0.03 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR