Diethyl ether ≥98.5% stabilised ACS for organic synthesis, for preparative liquid chromatography, general laboratory use

Supplier: BURDICK JACKSON
Danger


Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

LP108-4 LP108D19
BJAALP108-4EA 3940 SGD
BJAALP108-4 BJAALP108D19
Diethyl ether ≥98.5% stabilised ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Diethyl ether

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: (CH₃CH₂)₂O
Boiling Pt: 35 °C
Melting Pt: –117,4 °C
Density: 0,713 g/cm³ (20 °C)
Flash Pt: –45 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Alcohol (as CH3CH2OH) Passes test
Appearance Passes test
Assay by GC Analysis 98 % min.
Carbonyl (as HCHO) 0.001 % max.
Color (APHA) 10 max.
Peroxides (as H₂O₂) at time of packaging 1 ppm max.
Residue after Evaporation 0.001 % max.
Titrable Acid 0.2 µeq/g max.
Water by Karl Fischer Titration 0.03 % max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR