Hexane (mixture of isomers) 99,9 GC_ASSAY_METHOD (>60% n-Hexane), B&J Brand™, gradient HPLC_Pest_Residual, Spectrophotometry, Burdick & Jackson™

Supplier: BURDICK JACKSON
Danger

Synonyms: Hexanes, Isohexane

NP212-10 NP212-20 212-1 212-4
BJAANP212-10EA 846 SGD
BJAANP212-10 BJAANP212-20 BJAA212-1 BJAA212-4
Hexane (mixture of isomers) 99,9 GC_ASSAY_METHOD (>60% n-Hexane), B&J Brand™, gradient HPLC_Pest_Residual, Spectrophotometry, Burdick & Jackson™
Hexane (mixture of isomers)

Suitable for HPLC, gas chromatography, pesticide residue analysis, and spectrophotometry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68.7 °C
Melting Pt: –95 °C
Density: 0.659…0.673 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
GC Analysis (n-Hexane) 60 % min.
GC Analysis (total C6 isomers) 99.9 % min.
Residue 1 mg/L max.
UV Absorbance @ 225 nm 0.05 AU max.
UV Absorbance @ 250 nm 0.01 AU max.
UV Absorbance @ 275 nm 0.005 AU max.
UV Cutoff 195 nm max.
Water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR