Pyridine, anhydrous ≥99.8% for applications requiring low water content, Burdick & Jackson™
Supplier: BURDICK & JACKSON
|
Danger
|
AS332-0100
AS332-1
BJAAAS332-0100EA
1360
SGD
BJAAAS332-0100
BJAAAS332-1
Pyridine, anhydrous ≥99.8% for applications requiring low water content, Burdick & Jackson™
Pyridine
Septum-sealed bottle
- Water content ranging from <10 ppm to <50 ppm
- Low residue
- Convenient and effective packaging ensures product quality
- Certificate of Analysis printed on the label
Formula:
C₅H₅N Boiling Pt: 115,25 °C Melting Pt: –41,55 °C Density: 0,983 g/cm3 Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011732 CAS Number: 110-86-1 EINECS: 203-809-9 UN: 1282 ADR: 3,II Merck Index: 13,08060 |
Specification Test Results
Appearance | Passes test |
Color (APHA) | 10 max. |
GC Analysis | 99.8 % min. |
Residue | 5 mg/L max. |
Water by Karl Fischer Titration | 50 ppm max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...