Trifluoroacetic acid ≥99.5%, BioSyn™ for biosynthesis, for HPLC, Fluka™

Supplier: BURDICK JACKSON
Danger

BB360P050 BB360P500
BJAABB360P050EA 3440 SGD
BJAABB360P050 BJAABB360P500
Trifluoroacetic acid ≥99.5%, BioSyn™ for biosynthesis, for HPLC, Fluka™
Trifluoroacetic acid

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity

Formula: C₂HF₃O₂
MW: 114,02 g/mol
Boiling Pt: 72 °C
Melting Pt: –15,4 °C
Density: 1,48 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity 99.5 % min.
Ultraviolet Absorbance Passes test
Water by Karl Fischer Titration 0.1 % max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR