Water with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: BURDICK & JACKSON
NP486-20
486-4
BJAANP486-20EA
3060
SGD
BJAANP486-20
BJAA486-4
Water with 0,05% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,05% (v/v) trifluoroacetic acid
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Boiling Pt:
100 °C Melting Pt: 0 °C Density: 0,9982 g/cm³ (20 °C) Storage Temperature: Ambient |
EINECS:
200-929-3 |