Water with 0,1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: BURDICK JACKSON

LC485-1 LC485-2.5
BJAALC485-1EA 1420 SGD
BJAALC485-1 BJAALC485-2.5
Water with 0,1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Water with 0,1% (v/v) trifluoroacetic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0,9982 g/cm³ (20 °C)
Storage Temperature: Ambient
EINECS: 200-929-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR