Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Supplier: BURDICK & JACKSON
|
Danger
|
LC407-1
LC407-2.5
BJAALC407-1EA
1020
SGD
BJAALC407-1
BJAALC407-2.5
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, LabReady™, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Acetonitrile with 0.1% (v/v) trifluoroacetic acid
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Boiling Pt:
82 °C (1,013 hPa) Melting Pt: –46 °C Density: 0,777 g/cm³ (25 °C) Flash Pt: 6 °C (main component) Storage Temperature: Ambient |
EINECS:
200-929-3 UN: 1648 ADR: 3,II |