1,3-Dichloroacetone 99%
Supplier: Thermo Scientific
|
Danger
|
22302.06
22302.14
22302.22
ALFA22302.06EA
0
SGD
ALFA22302.06
ALFA22302.14
ALFA22302.22
1,3-Dichloroacetone 99%
1,3-Dichloroacetone
Purity: 99%
CAS No.: 534-07-6
Molecular Formula: ClCH2C(=O)CH2Cl
Formula Weight: 126.97
Flash Point: 89°(193°F)
Physical Form: Crystalline
Density: 1.3834
Refractive Index: 1.4400
Odour: Odourless
Melting Point: 43-45°
Boiling Point: 173°
Beilstein Reference: 605456
Merck Reference: 14,3052
UN No.: 2649
MDL No.: MFCD00000937
Formula:
ClH₂CC(O)CH₂Cl MW: 126,97 g/mol Boiling Pt: 173 °C Melting Pt: 43…45 °C Density: 1,3834 Flash Pt: 89 °C (193 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000937 CAS Number: 534-07-6 EINECS: 208-585-6 UN: 2649 ADR: 6.1,II Merck Index: 14,3052 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...