Hexachloroacetone ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Hexachloro-2-propanone
B23259.0B
B23259.22
B23259.30
ALFAB23259.0BEA
279
SGD
ALFAB23259.0B
ALFAB23259.22
ALFAB23259.30
Hexachloroacetone ≥99%
Hexachloroacetone
Purity: 99%
CAS No.: 116-16-5
Molecular Formula: C3Cl6O
Formula Weight: 264.75
Flash Point:
Formula:
C₃Cl₆O MW: 264,75 g/mol Boiling Pt: 203…205 °C Melting Pt: –2 °C Density: 1,748 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000796 CAS Number: 116-16-5 EINECS: 204-129-5 UN: 2661 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...