1,1,1-Trifluoroacetone 95%

Supplier: Thermo Scientific
Danger

A13556.14 A13556.22
ALFAA13556.14EA 0 SGD
ALFAA13556.14 ALFAA13556.22
1,1,1-Trifluoroacetone 95%
1,1,1-Trifluoroacetone

Purity: 95% CAS No.: 421-50-1 Molecular Formula: CF3COCH3 Formula Weight: 112.05 Storage Temperature: 2°C to 8°C Flash Point: -31°(-24°F) Refractive Index: 1.3000 Odour: Chloroform-like Melting Point: -78° Boiling Point: 21-24° Beilstein Reference: 1748614 UN No.: 1993 MDL No.: MFCD00000423

Formula: C₃H₃F₃O
Boiling Pt: 21…24 °C
Melting Pt: < –78 °C
Density: 1,252
Flash Pt: –31 °C (–24 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000423
CAS Number: 421-50-1
EINECS: 207-005-9
UN: 1993
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR