1,1,1,2,2-Pentafluoro-3-iodopropane 96%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Iodo-1,1,1,2,2-pentafluoropropane

L11047.03 L11047.06
ALFAL11047.03EA 87.7 SGD
ALFAL11047.03 ALFAL11047.06
1,1,1,2,2-Pentafluoro-3-iodopropane 96%
1,1,1,2,2-Pentafluoro-3-iodopropane

Purity: 96% CAS No.: 354-69-8 Molecular Formula: CF3CF2CH2I Formula Weight: 259.95 Refractive Index: 1.3730 Boiling Point: 70-71° MDL No.: MFCD00039405

Formula: C₃H₂F₅I
MW: 259,95 g/mol
Boiling Pt: 70…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00039405
CAS Number: 354-69-8
EINECS: 206-568-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR