Pentyl isocyanate ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 1-Pentyl isocyanate

L13399.03 L13399.06
ALFAL13399.03EA 85.2 SGD
ALFAL13399.03 ALFAL13399.06
Pentyl isocyanate ≥97%
Pentyl isocyanate
Formula: C₆H₁₁NO
MW: 113,16 g/mol
Boiling Pt: 136…137 °C
Density: 0,878
Flash Pt: 32 °C (89 °F)
MDL Number: MFCD00144846
CAS Number: 3954-13-0
UN: 3080
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR