1-Bromo-4,4,4-trifluorobutane ≥98%

Supplier: Thermo Fisher Scientific
Warning

L10237.03 L10237.06
ALFAL10237.03EA 80.3 SGD
ALFAL10237.03 ALFAL10237.06
1-Bromo-4,4,4-trifluorobutane ≥98%
1-Bromo-4,4,4-trifluorobutane

Purity: 98% CAS No.: 406-81-5 Molecular Formula: CF3CH2CH2CH2Br Formula Weight: 191.00 Refractive Index: 1.3820 Boiling Point: 103-105° MDL No.: MFCD00066335

Formula: C₄H₆BrF₃
MW: 190.99 g/mol
Boiling Pt: 103…105 °C
Density: 1.555
Storage Temperature: Ambient
MDL Number: MFCD00066335
CAS Number: 406-81-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR