1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol tech. 90%, Technical Grade

Supplier: Thermo Scientific

Synonyms: 1H,1H,12H,12H-Perfluoro-1,12-dodecanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Eicosafluoro-1,12-dodecandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11-Icosafluorododecane-1,12-diol, 1H,1H,12H,12H-Icosafluoro-1,12-dodecanediol, 1H,1H,12H,12H-Perfluoro-1,12-dodecandiol

L16588.03 L16588.06
ALFAL16588.03EA 0 SGD
ALFAL16588.03 ALFAL16588.06
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol tech. 90%, Technical Grade
1H,1H,12H,12H-Eicosafluoro-1,12-dodecandiol

Purity: 90% CAS No.: 183162-43-8 Molecular Formula: HOCH2(CF2)10CH2OH Formula Weight: 562.15 Melting Point: 156-160° Boiling Point: 170°/5mm MDL No.: MFCD00236625

Formula: C₁₂H₆F₂₀O₂
MW: 562,15 g/mol
Boiling Pt: 170 °C (5 mmHg)
Melting Pt: 156…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00236625
CAS Number: 183162-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR