1H,1H,9H,9H-Perfluoro-1,9-nonanediol tech. 90%

Supplier: Thermo Scientific

Synonyms: 1H,1H,9H,9H-Perfluoro-1,9-nonanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonandiol

L16599.06 L16599.14
ALFAL16599.06EA 149 SGD
ALFAL16599.06 ALFAL16599.14
1H,1H,9H,9H-Perfluoro-1,9-nonanediol tech. 90%
1H,1H,9H,9H-Tetradecafluoro-1,9-nonandiol

Purity: 90% CAS No.: 203303-01-9 Molecular Formula: C9H6F14O2 Formula Weight: 412.12 Melting Point: 108-112° Boiling Point: 140°/5mm MDL No.: MFCD00153275

Formula: C₉H₆F₁₄O₂
MW: 412,12 g/mol
Boiling Pt: 140 °C (5 mmHg)
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00153275
CAS Number: 203303-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR