1H,1H-Heptacosafluoro-1-tetradecanol 96%
Supplier: Thermo Scientific
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluoro-1-tetradecanol, 1H,1H-Perfluoro-1-tetradecanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluor-1-tetradecanol
L16609.06
L16609.14
ALFAL16609.06EA
195
SGD
ALFAL16609.06
ALFAL16609.14
1H,1H-Heptacosafluoro-1-tetradecanol 96%
1H,1H-Heptacosafluoro-1-tetradecanol
Purity: 96%
CAS No.: 15622-57-8
Molecular Formula: CF3(CF2)12CH2OH
Formula Weight: 700.14
Melting Point: 137-140°
Boiling Point: 130°/60mm
MDL No.: MFCD00153264
Formula:
C₁₄H₃F₂₇O MW: 700,13 g/mol Boiling Pt: 130 °C (60 mmHg) Melting Pt: 137…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153264 CAS Number: 15622-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...