3,3-Bis(bromomethyl)pentane tech. 90%, Technical Grade

Supplier: Thermo Scientific
Warning

Synonyms: 1,3-Dibromo-2,2-diethylpropane

L09629.09 L09629.18
ALFAL09629.09EA 0 SGD
ALFAL09629.09 ALFAL09629.18
3,3-Bis(bromomethyl)pentane tech. 90%, Technical Grade
3,3-Bis(bromomethyl)pentane

Purity: 90% CAS No.: 67969-84-0 Molecular Formula: (CH3CH2)2C(CH2Br)2 Formula Weight: 258.01 Melting Point: 38-41° Boiling Point: 96-97°/10mm MDL No.: MFCD00039190

Formula: C₇H₁₄Br₂
MW: 258 g/mol
Boiling Pt: 96…97 °C (10 mmHg)
Melting Pt: 38…41 °C
Storage Temperature: Ambient
MDL Number: MFCD00039190
CAS Number: 67969-84-0
EINECS: 267-985-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR