9,10-Dibromoanthracene ≥98%
Supplier: Thermo Scientific
|
Warning
|
L12680.14
L12680.06
L12680.22
ALFAL12680.14EA
295
SGD
ALFAL12680.14
ALFAL12680.06
ALFAL12680.22
9,10-Dibromoanthracene ≥98%
9,10-Dibromoanthracene
Purity: 98%
CAS No.: 523-27-3
Molecular Formula: C14H8Br2
Formula Weight: 336.04
Melting Point: 222-223° subl.
Beilstein Reference: 1912109
Merck Reference: 14,3018
MDL No.: MFCD00001244
Formula:
C₁₄H₈Br₂ MW: 336,03 g/mol Boiling Pt: 427 °C (1013 hPa) Melting Pt: 222…223 °C (subl.) Density: 1,981 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001244 CAS Number: 523-27-3 EINECS: 208-342-4 Merck Index: 14,3018 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...