2,4-Dibromo-1-fluorobenzene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1,3-Dibromo-4-fluorobenzene, 1-Fluoro-2,4-dibromobenzene

L20171.06 L20171.14
ALFAL20171.06EA 0 SGD
ALFAL20171.06 ALFAL20171.14
2,4-Dibromo-1-fluorobenzene ≥98%
2,4-Dibromo-1-fluorobenzene

Purity: 98% CAS No.: 1435-53-6 Molecular Formula: C6H3Br2F Formula Weight: 253.91 Flash Point: 92°(197°F) Refractive Index: 1.5840 Boiling Point: 103°/32mm Beilstein Reference: 1934822 MDL No.: MFCD00000283

Formula: C₆H₃Br₂F
MW: 253,9 g/mol
Boiling Pt: 103 °C (32 mmHg)
Density: 2,047
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000283
CAS Number: 1435-53-6
EINECS: 215-860-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR