Ethyl-O-anisate ≥98%

Supplier: Thermo Scientific

Synonyms: 2-Methoxybenzoic acid ethyl ester, Ethyl 2-methoxybenzoate, o-Anisic acid ethyl ester, 2-Anisic acid ethyl ester

A11287.09 A11287.18
ALFAA11287.09EA 0 SGD
ALFAA11287.09 ALFAA11287.18
Ethyl-O-anisate ≥98%
Ethyl o-anisate

Purity: 98+% CAS No.: 7335-26-4 Molecular Formula: C10H12O3 Formula Weight: 180.20 Refractive Index: 1.5210 Boiling Point: 127-128°/11mm MDL No.: MFCD00015148

Formula: C₁₀H₁₂O₃
MW: 180,2 g/mol
Boiling Pt: 127…128 °C (11 mmHg)
Density: 1,112
Storage Temperature: Ambient
MDL Number: MFCD00015148
CAS Number: 7335-26-4
EINECS: 230-843-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR