2-Vinylnaphthalene ≥97% stabilised

Supplier: Thermo Scientific
Warning


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A12470.06 A12470.14
ALFAA12470.06EA 147 SGD
ALFAA12470.06 ALFAA12470.14
2-Vinylnaphthalene ≥97% stabilised
2-Vinylnaphthalene

Purity: 97% CAS No.: 827-54-3 Molecular Formula: C12H10 Formula Weight: 154.21 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Melting Point: 62-67° Boiling Point: 128-131°/13mm Beilstein Reference: 2038190 MDL No.: MFCD00004125

Formula: C₁₂H₁₀
MW: 154,21 g/mol
Boiling Pt: 128…131 °C (13 mmHg)
Melting Pt: 62…67 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00004125
CAS Number: 827-54-3
EINECS: 212-573-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR