Ethyl-3-nitrocinnamate ≥98%

Supplier: Thermo Scientific

Synonyms: Ethyl 3-(3-nitrophenyl)acrylate

A12995.06 A12995.14
ALFAA12995.06EA 0 SGD
ALFAA12995.06 ALFAA12995.14
Ethyl-3-nitrocinnamate ≥98%
Ethyl 3-nitrocinnamate

Purity: 98% CAS No.: 5396-71-4 Molecular Formula: C11H11NO4 Formula Weight: 221.21 Melting Point: 74-76° Beilstein Reference: 2618102

Formula: C₁₁H₁₁NO₄
MW: 221,21 g/mol
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00017027
CAS Number: 5396-71-4
EINECS: 226-415-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR