Ethyl-4-nitrocinnamate ≥99%
Supplier: Thermo Scientific
Synonyms:
Ethyl 4-nitrocinnamate
A13196.06
A13196.14
A13196.22
ALFAA13196.06EA
0
SGD
ALFAA13196.06
ALFAA13196.14
ALFAA13196.22
Ethyl-4-nitrocinnamate ≥99%
Ethyl 4-nitrocinnamate
Purity: 99%
CAS No.: 953-26-4
Molecular Formula: C11H11NO4
Formula Weight: 221.21
Melting Point: 138-140°
Beilstein Reference: 2052533
Formula:
C₁₁H₁₁NO₄ MW: 221,21 g/mol Melting Pt: 138…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007380 CAS Number: 953-26-4 EINECS: 213-463-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...