Phenylsulphonylacetone ≥97%
Supplier: Thermo Scientific
Synonyms:
1-(Phenylsulphonyl)acetone, (2-Propanoylsulphonyl)benzol, 1-(Phenylsulphonyl)propan-2-one, Benzenesulphonylacetone
A14085.03
A14085.06
A14085.14
ALFAA14085.03EA
0
SGD
ALFAA14085.03
ALFAA14085.06
ALFAA14085.14
Phenylsulphonylacetone ≥97%
Phenylsulphonylacetone
Purity: 97%
CAS No.: 5000-44-2
Molecular Formula: C9H10O3S
Formula Weight: 198.24
Melting Point: 54-58°
Beilstein Reference: 640928
Formula:
C₉H₁₀O₃S MW: 198,24 g/mol Melting Pt: 54…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047794 CAS Number: 5000-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...